Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181579.1 | NP_853557.1 | 12 - 13 | 1431 | 140 | |
GenBank mRNA | AF309775.1 | - | 11 - 12 | 1324 | 140 | |
AK036922.1 | - | 12 - 13 | 1438 | 140 | ||
AK137653.1 | - | 12 - 13 | 1430 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |