Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400009573.mm.1 |
NM_175500.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009573.mm.1 |
NM_175500.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009573.mm.1 |
TC1400001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009575.mm.1 |
NM_175500.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009575.mm.1 |
NM_175500.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009575.mm.1 |
TC1400001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009576.mm.1 |
NM_175500.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009576.mm.1 |
NM_175500.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009576.mm.1 |
TC1400001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009571.mm.1 |
NM_175500.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009571.mm.1 |
NM_175500.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009571.mm.1 |
TC1400001247.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009574.mm.1 |
NM_175500.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009574.mm.1 |
NM_175500.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009574.mm.1 |
TC1400001247.mm.1 |
MTA TC |