Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080699.1 | - | 171 | 65 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 15386 | Gm15386 |
BD-17, Defb17, OTTMUSG00000020946, OTTMUSG00000020948 |
-810.0kb | Sense | Chr. 1 - 18260524 - 18265138 on NCBI Build GRCm38 |
Downstream | transcription factor AP-2 beta | Tfap2b |
AI606113, AP-2(beta), E130018K07Rik, Tcfap2b |
104.1kb | Anti-sense | Chr. 1 - 19208914 - 19238845 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons |