Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134117.2 | NP_598878.2 | 2 - 3 | 943 | 119 | |
GenBank mRNA | AB381893.1 | - | 2 - 3 | 935 | 119 | |
AB436780.1 | - | 2 - 3 | 924 | 119 | ||
BC022157.1 | - | 1 - 2 | 90 | 119 | ||
BC086639.1 | - | 3 - 4 | 781 | 6828629 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |