Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163622.1 | NP_001157094.1 | 4 - 5 | 887 | 132 | |
NM_001163623.1 | NP_001157095.1 | 3 - 4 | 351 | 132 | ||
NM_001163624.1 | NP_001157096.1 | 3 - 4 | 219 | 132 | ||
NM_145984.3 | NP_666096.3 | 4 - 5 | 451 | 132 | ||
GenBank mRNA | AK028877.1 | - | 4 - 5 | 845 | 132 | |
AK050878.1 | - | 3 - 4 | 213 | 132 | ||
AK083070.1 | - | 4 - 5 | 392 | 132 | ||
AK163108.1 | - | 4 - 5 | 852 | 132 | ||
BC004612.1 | - | 3 - 4 | 305 | 3585402 | 132 | |
BC129880.1 | - | 2 - 3 | 883 | 132 | ||
BY257815.1 | - | 3 - 4 | 220 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |