Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159496.1 | NP_001152968.1 | 5 - 6 | 1542 | 89 | |
GenBank mRNA | AJ271833.1 | - | 5 - 6 | 1520 | 89 | |
AK010224.1 | - | 4 - 5 | 445 | 89 | ||
AK136077.1 | - | 5 - 6 | 1544 | 89 | ||
AK160653.1 | - | 5 - 6 | 1543 | 89 | ||
AK167176.1 | - | 5 - 6 | 1541 | 89 | ||
BC018556.1 | - | 5 - 6 | 1233 | 3987825 | 89 | |
BC090963.1 | - | 4 - 5 | 1133 | 30632168 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |