Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4htm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028944.3 | NP_083220.3 | 1 - 2 | 389 | 76 | |
GenBank mRNA | AK016685.1 | - | 1 - 2 | 363 | 76 | |
AK038927.1 | - | 1 - 2 | 371 | 76 | ||
AK046783.1 | - | 1 - 2 | 401 | 76 | ||
AK047714.1 | - | 1 - 2 | 389 | 76 | ||
AK161992.1 | - | 1 - 2 | 365 | 76 | ||
BC111871.1 | - | 1 - 2 | 389 | 40057593 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |