Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026378.3 | NP_080654.3 | 1 - 2 | 186 | 67 | |
GenBank mRNA | AK149963.1 | - | 1 - 2 | 187 | 67 | |
AK169405.1 | - | 1 - 2 | 184 | 67 | ||
AK171818.1 | - | 1 - 2 | 187 | 67 | ||
BC013483.1 | - | 1 - 2 | 167 | 4242135 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |