Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144860.2 | NP_659109.2 | 10 - 11 | 2154 | 94 | |
GenBank mRNA | AK053035.1 | - | 3 - 4 | 377 | 94 | |
AK080847.1 | - | 4 - 5 | 430 | 94 | ||
AY147848.1 | - | 10 - 11 | 1653 | 94 | ||
AY149907.1 | - | 10 - 11 | 1474 | 94 | ||
AY245539.1 | - | 10 - 11 | 2154 | 94 | ||
AY974091.1 | - | 10 - 11 | 1447 | 94 | ||
BC131662.1 | - | 11 - 12 | 1620 | 40110696 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |