Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010258.3 | NP_034388.2 | 4 - 5 | 1683 | 83 | |
GenBank mRNA | AF179425.1 | - | 3 - 4 | 990 | 83 | |
AK142381.1 | - | 4 - 5 | 1543 | 83 | ||
S82462.1 | - | 3 - 4 | 1445 | 83 | ||
U51335.1 | - | 2 - 3 | 263 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |