Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285940.1 | NP_001272869.1 | 4 - 5 | 688 | 113 | |
NM_001285946.1 | NP_001272875.1 | 4 - 5 | 688 | 113 | ||
NM_001285949.1 | NP_001272878.1 | 4 - 5 | 688 | 113 | ||
NM_199448.3 | NP_955519.1 | 4 - 5 | 688 | 113 | ||
GenBank mRNA | AK033547.1 | - | 4 - 5 | 630 | 113 | |
AK150241.1 | - | 4 - 5 | 654 | 113 | ||
AK157221.1 | - | 4 - 5 | 655 | 113 | ||
AK159545.1 | - | 4 - 5 | 641 | 113 | ||
AK165595.1 | - | 4 - 5 | 638 | 113 | ||
AY382580.1 | - | 4 - 5 | 639 | 113 | ||
BC052366.1 | - | 4 - 5 | 667 | 113 | ||
BC064130.1 | - | 4 - 5 | 594 | 113 | ||
BC096619.1 | - | 4 - 5 | 633 | 6391451 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |