Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028719.1 | NP_082995.1 | 2 - 3 | 499 | 65 | |
GenBank mRNA | AK014396.1 | - | 2 - 3 | 501 | 65 | |
AK016705.1 | - | 5 - 6 | 1034 | 65 | ||
AK043692.1 | - | 2 - 3 | 512 | 65 | ||
BC043087.1 | - | 2 - 3 | 499 | 6417611 | 65 | |
BC063081.1 | - | 2 - 3 | 499 | 6417611 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |