Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177775.3 | NP_808443.2 | 8 - 9 | 1069 | 72 | |
GenBank mRNA | AK086367.1 | - | 8 - 9 | 1070 | 72 | |
AK220532.1 | - | 8 - 9 | 1057 | 72 | ||
BC118967.2 | - | 1017 | 40097262 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |