Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276442.1 | NP_001263371.1 | 8 - 9 | 73 | ||
NM_025371.3 | NP_079647.1 | 8 - 9 | 73 | |||
GenBank mRNA | AK002423.1 | - | 8 - 9 | 73 | ||
AK003703.1 | - | 8 - 9 | 73 | |||
BC005631.1 | - | 8 - 9 | 701 | 3964138 | 73 | |
CV560337.1 | - | 8 - 9 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |