Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291835.1 | NP_001278764.1 | 4 - 5 | 1159 | 147 | |
NM_020559.2 | NP_065584.2 | 4 - 5 | 772 | 147 | ||
GenBank mRNA | AK077152.1 | - | 3 - 4 | 463 | 147 | |
AK133283.1 | - | 4 - 5 | 1160 | 147 | ||
BC022110.1 | - | 4 - 5 | 5133249 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |