Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026497.2 | NP_080773.1 | 1 - 2 | 102 | 109 | |
GenBank mRNA | AK002641.1 | - | 1 - 2 | 103 | 109 | |
AK083846.1 | - | 1 - 2 | 60 | 109 | ||
AK147867.1 | - | 1 - 2 | 56 | 109 | ||
BC057657.1 | - | 1 - 2 | 56 | 2646355 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |