Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026497.2 | NP_080773.1 | 4 - 5 | 1080 | 86 | |
GenBank mRNA | AK002641.1 | - | 4 - 5 | 1081 | 86 | |
AK083846.1 | - | 4 - 5 | 1038 | 86 | ||
AK147867.1 | - | 4 - 5 | 1034 | 86 | ||
BC057657.1 | - | 4 - 5 | 1034 | 2646355 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |