Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018818.2 | NP_061288.2 | 3 - 4 | 212 | 81 | |
GenBank mRNA | AF218084.1 | - | 3 - 4 | 212 | 81 | |
AK035956.1 | - | 3 - 4 | 207 | 81 | ||
AK141830.1 | - | 3 - 4 | 207 | 81 | ||
BC047461.1 | - | 3 - 4 | 202 | 5067077 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |