Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012021.2 | NP_036151.1 | 3 - 4 | 455 | 110 | |
GenBank mRNA | AF110733.1 | - | 3 - 4 | 381 | 110 | |
AF124994.1 | - | 3 - 4 | 544 | 110 | ||
AF197951.1 | - | 3 - 4 | 455 | 110 | ||
AK002383.1 | - | 3 - 4 | 421 | 110 | ||
AK003332.2 | - | 3 - 4 | 421 | 110 | ||
AK009858.1 | - | 3 - 4 | 890 | 110 | ||
AK136130.1 | - | 3 - 4 | 396 | 110 | ||
AK152653.1 | - | 3 - 4 | 421 | 110 | ||
BC008174.1 | - | 3 - 4 | 840 | 3481713 | 110 | |
BC014682.1 | - | 1 - 2 | 65 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |