Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144532.2 | NP_653115.1 | 3 - 4 | 579 | 82 | |
GenBank mRNA | AY039218.1 | - | 3 - 4 | 574 | 82 | |
BC049263.1 | - | 3 - 4 | 579 | 5366999 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |