Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178650.3 | NP_848765.2 | 5 - 6 | 517 | 75 | |
GenBank mRNA | AK040487.1 | - | 5 - 6 | 518 | 75 | |
AK075803.1 | - | 5 - 6 | 504 | 75 | ||
AK131177.1 | - | 4 - 5 | 498 | 75 | ||
AK146426.1 | - | 5 - 6 | 511 | 75 | ||
AK153836.1 | - | 5 - 6 | 515 | 75 | ||
AK154444.1 | - | 4 - 5 | 495 | 75 | ||
BC094901.1 | - | 4 - 5 | 492 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |