Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178650.3 | NP_848765.2 | 6 - 7 | 632 | 63 | |
GenBank mRNA | AK040487.1 | - | 6 - 7 | 633 | 63 | |
AK075803.1 | - | 6 - 7 | 619 | 63 | ||
AK131177.1 | - | 5 - 6 | 613 | 63 | ||
AK146426.1 | - | 6 - 7 | 626 | 63 | ||
AK153836.1 | - | 6 - 7 | 630 | 63 | ||
BC094901.1 | - | 5 - 6 | 607 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |