Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081080.1 | NP_001074549.1 | 10 - 11 | 3329 | 101 | |
GenBank mRNA | AK122230.1 | - | 6 - 7 | 780 | 101 | |
BC099524.1 | - | 4 - 5 | 412 | 101 | ||
BC137913.1 | - | 10 - 11 | 3335 | 8860932 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |