Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081309.1 | NP_001074778.1 | 14 - 15 | 3698 | 77 | |
GenBank mRNA | AK050333.1 | - | 14 - 15 | 3668 | 77 | |
AK054559.1 | - | 14 - 15 | 3715 | 77 | ||
AK087938.1 | - | 14 - 15 | 3689 | 77 | ||
BC023047.1 | - | 3 - 4 | 347 | 77 | ||
BC024083.1 | - | 11 - 12 | 2172 | 77 | ||
BC157947.1 | - | 14 - 15 | 3523 | 9007281 | 77 | |
BC158047.1 | - | 14 - 15 | 3523 | 9088160 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |