Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110791.1 | NP_001104261.1 | 10 - 11 | 1747 | 63 | |
NM_011750.2 | NP_035880.2 | 10 - 11 | 1747 | 63 | ||
GenBank mRNA | AK146151.1 | - | 10 - 11 | 1508 | 63 | |
AK147081.1 | - | 10 - 11 | 1516 | 63 | ||
AK154705.1 | - | 10 - 11 | 1515 | 63 | ||
AK157845.1 | - | 10 - 11 | 1516 | 63 | ||
AK194686.1 | - | 1 - 2 | 259 | 63 | ||
BC009091.1 | - | 10 - 11 | 1492 | 3157495 | 63 | |
BC055370.1 | - | 10 - 11 | 1506 | 30055109 | 63 | |
X80159.1 | - | 2 - 3 | 464 | 63 | ||
X85802.1 | - | 10 - 11 | 1504 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |