Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205234.1 | NP_001192163.1 | 13 - 14 | 2997 | 75 | |
NM_001205235.1 | NP_001192164.1 | 12 - 13 | 2973 | 75 | ||
NM_020253.3 | NP_064649.2 | 13 - 14 | 2997 | 75 | ||
GenBank mRNA | AK129239.1 | - | 13 - 14 | 2970 | 75 | |
AK147295.1 | - | 13 - 14 | 2933 | 75 | ||
AK147665.1 | - | 13 - 14 | 2932 | 75 | ||
BC145496.1 | - | 11 - 12 | 2620 | 9054059 | 75 | |
BC150784.1 | - | 11 - 12 | 2620 | 9087695 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |