Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028782.2 | NP_083058.2 | 7 - 8 | 1143 | 67 | |
GenBank mRNA | AK131910.1 | - | 7 - 8 | 1129 | 67 | |
AK157474.1 | - | 7 - 8 | 1122 | 67 | ||
AK159302.1 | - | 7 - 8 | 1128 | 67 | ||
AK160071.1 | - | 7 - 8 | 1128 | 67 | ||
AK160076.1 | - | 7 - 8 | 1129 | 67 | ||
AK161983.1 | - | 7 - 8 | 1128 | 67 | ||
AK201275.1 | - | 1 - 2 | 75 | 67 | ||
AK205545.1 | - | 1 - 2 | 75 | 67 | ||
AY190302.1 | - | 7 - 8 | 1113 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |