Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142631.1 | NP_001136103.1 | 2 - 3 | 319 | 79 | |
NM_029254.1 | NP_083530.1 | 2 - 3 | 319 | 79 | ||
GenBank mRNA | AK014910.1 | - | 2 - 3 | 309 | 79 | |
AK077138.1 | - | 2 - 3 | 320 | 79 | ||
AY820306.1 | - | 1 - 2 | 220 | 79 | ||
AY820307.1 | - | 1 - 2 | 220 | 79 | ||
BC147706.1 | - | 2 - 3 | 297 | 9087465 | 79 | |
BC147719.1 | - | 2 - 3 | 297 | 9087478 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |