Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335318 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335314 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335313 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335311 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335315 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335316 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335317 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335320 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954156 |
AK152321.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954156 |
BC079913.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954156 |
AK032907.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954156 |
AK129031.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1954156 |
NM_175639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
118912 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
221894 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683742 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
206781 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533964 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696439 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
965579 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838812 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537771 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
145347 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
845006 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
383139 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562737 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
73003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437659 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521104 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492792 |
AK009469.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492792 |
AK158031.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492792 |
BC016454.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492792 |
NM_027984.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
847928 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
787615 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397895 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376407 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
103753 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
337357 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
434801 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
657038 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
372128 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
72494 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83627 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
177841 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
7986 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P113907 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110336 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954156 |
AK152321.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954156 |
BC079913.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954156 |
AK032907.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954156 |
AK129031.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1954156 |
NM_175639.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08844735 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110336 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08844803 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P113907 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P8174 |
BC117724.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P8174 |
AK152321.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P8174 |
BC079913.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P8174 |
AK032907.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P8174 |
AK129031.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
BC079913.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
AK129031.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P384901 |
NM_175639.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P111561 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110336 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P113907 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335320 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335318 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335314 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335311 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335316 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335317 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335315 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31335313 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335324 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335345 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335332 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335458 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13224794 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335489 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335360 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335334 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335348 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335415 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335353 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13240905 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335482 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335318 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335412 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335302 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13304038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335468 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335330 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335410 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335467 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13223618 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13224803 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335338 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335480 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13091106 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335359 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335473 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335352 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335339 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335328 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335301 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335342 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335333 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335355 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335305 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335326 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13091113 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335298 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13226472 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335351 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335462 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335347 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335314 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335419 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335472 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335336 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335413 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13558815 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335362 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335294 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335470 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335296 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13261185 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335310 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335331 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13223621 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335420 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335343 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335350 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335299 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335466 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13240911 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335479 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13304036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335474 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335469 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335340 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335460 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335484 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335341 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335411 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31335291 |
- |
- |