Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025602.3 | NP_001020773.1 | 1 - 2 | 227 | 71 | |
NM_010743.3 | NP_034873.2 | 1 - 2 | 227 | 71 | ||
GenBank mRNA | AK033417.1 | - | 1 - 2 | 80 | 71 | |
M24843.1 | - | 1 - 2 | 68 | 71 | ||
Y07519.1 | - | 1 - 2 | 71 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |