Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011283.2 | NP_035413.1 | 1 - 2 | 117 | 122 | |
GenBank mRNA | AK044413.1 | - | 1 - 2 | 110 | 122 | |
BC096607.1 | - | 1 - 2 | 117 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |