Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700023077.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023077.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023077.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023081.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023081.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023081.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023078.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023078.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023078.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023080.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023080.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023080.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023083.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023083.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023083.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023079.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023079.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023079.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023084.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023084.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023084.mm.1 |
TC1700002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023082.mm.1 |
NM_001033444.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023082.mm.1 |
NM_001033444.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023082.mm.1 |
TC1700002519.mm.1 |
MTA TC |