Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010109.3 | NP_034239.1 | 1 - 2 | 309 | 94 | |
NM_207654.2 | NP_997537.1 | 1 - 2 | 309 | 94 | ||
GenBank mRNA | BC040218.1 | - | 1 - 2 | 309 | 4954352 | 94 |
U90664.1 | - | 1 - 2 | 136 | 94 | ||
U90665.1 | - | 1 - 2 | 136 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |