Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042607.1 | NP_001036072.1 | 11 - 12 | 1398 | 66 | |
NM_001284258.1 | NP_001271187.1 | 10 - 11 | 1276 | 66 | ||
NM_013649.3 | NP_038677.3 | 11 - 12 | 1407 | 66 | ||
GenBank mRNA | AK076449.1 | - | 5 - 6 | 500 | 66 | |
AK144355.1 | - | 10 - 11 | 1216 | 66 | ||
BC006963.1 | - | 7 - 8 | 706 | 66 | ||
BC032275.1 | - | 11 - 12 | 1132 | 5370100 | 66 | |
L02210.1 | - | 11 - 12 | 1382 | 66 | ||
L21707.1 | - | 11 - 12 | 1416 | 66 | ||
L38394.1 | - | 11 - 12 | 1330 | 66 | ||
M98547.1 | - | 11 - 12 | 1371 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |