Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042607.1 | NP_001036072.1 | 12 - 13 | 1502 | 71 | |
NM_001284258.1 | NP_001271187.1 | 11 - 12 | 1380 | 71 | ||
NM_013649.3 | NP_038677.3 | 12 - 13 | 1511 | 71 | ||
GenBank mRNA | AK076449.1 | - | 6 - 7 | 604 | 71 | |
AK144355.1 | - | 11 - 12 | 1320 | 71 | ||
BC006963.1 | - | 8 - 9 | 810 | 71 | ||
BC032275.1 | - | 12 - 13 | 1236 | 5370100 | 71 | |
L02210.1 | - | 12 - 13 | 1486 | 71 | ||
L21707.1 | - | 12 - 13 | 1520 | 71 | ||
L38394.1 | - | 12 - 13 | 1434 | 71 | ||
M98547.1 | - | 12 - 13 | 1475 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |