Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027974.1 | NP_082250.1 | 8 - 9 | 1607 | 131 | |
GenBank mRNA | AK006489.1 | - | 8 - 9 | 1546 | 131 | |
EU520262.1 | - | 8 - 9 | 1499 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |