Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009573.3 | NP_033599.2 | 1 - 2 | 1759 | 80 | |
GenBank mRNA | AK136977.1 | - | 1 - 2 | 984 | 80 | |
AK141883.1 | - | 1 - 2 | 953 | 80 | ||
AK159769.1 | - | 1 - 2 | 1731 | 80 | ||
BC050889.1 | - | 1 - 2 | 997 | 6418777 | 80 | |
BC060247.1 | - | 2 - 3 | 1339 | 6813617 | 80 | |
BC063247.1 | - | 1 - 2 | 1721 | 5694793 | 80 | |
D32167.1 | - | 1 - 2 | 1507 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |