Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 16 - 17 | 4016 | 60 | |
GenBank mRNA | AK147671.1 | - | 16 - 17 | 4032 | 60 | |
AK172931.1 | - | 6 - 7 | 2325 | 60 | ||
BC079860.1 | - | 16 - 17 | 4016 | 6834745 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |