Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134164.5 | NP_598925.1 | 2 - 3 | 262 | 79 | |
GenBank mRNA | AB062804.1 | - | 1 - 2 | 30 | 79 | |
AK046627.1 | - | 2 - 3 | 566 | 79 | ||
AK083139.1 | - | 2 - 3 | 263 | 79 | ||
BC116846.1 | - | 1 - 2 | 60 | 40126165 | 79 | |
BC116848.1 | - | 1 - 2 | 60 | 40126167 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |