Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013456.2 | NP_038484.1 | 7 - 8 | 763 | 106 | |
GenBank mRNA | AF093775.1 | - | 7 - 8 | 738 | 106 | |
BC111890.1 | - | 7 - 8 | 729 | 40060598 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |