Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001984.2 | NP_001001984.2 | 18 - 19 | 3759 | 109 | |
GenBank mRNA | AK141779.1 | - | 4 - 5 | 1000 | 109 | |
AK173084.1 | - | 7 - 8 | 1577 | 109 | ||
BC036264.1 | - | 6 - 7 | 1458 | 109 | ||
BC037117.1 | - | 11 - 12 | 2308 | 109 | ||
BC057051.1 | - | 5 - 6 | 1132 | 109 | ||
BC076576.1 | - | 18 - 19 | 3324 | 6840591 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |