Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001984.2 | NP_001001984.2 | 9 - 10 | 1670 | 148 | |
GenBank mRNA | AK037157.1 | - | 10 - 11 | 1567 | 148 | |
AK155866.1 | - | 9 - 10 | 1675 | 148 | ||
BC037117.1 | - | 2 - 3 | 219 | 148 | ||
BC076576.1 | - | 9 - 10 | 1236 | 6840591 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |