Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290818.1 | NP_001277747.1 | 7 - 8 | 766 | 72 | |
NM_130863.2 | NP_570933.1 | 8 - 9 | 838 | 72 | ||
GenBank mRNA | AF333028.1 | - | 7 - 8 | 558 | 72 | |
AK155691.1 | - | 7 - 8 | 767 | 72 | ||
AK193363.1 | - | 2 - 3 | 79 | 72 | ||
BC003196.1 | - | 7 - 8 | 554 | 72 | ||
BC053922.1 | - | 8 - 9 | 571 | 6309555 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |