Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 10 - 11 | 981 | 89 | |
GenBank mRNA | AK037753.1 | - | 10 - 11 | 967 | 89 | |
AK045850.1 | - | 10 - 11 | 979 | 89 | ||
AK163815.1 | - | 1 - 2 | 149 | 89 | ||
BC076577.1 | - | 10 - 11 | 970 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |