Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102423.1 | NP_001095893.1 | 5 - 6 | 647 | 82 | |
NM_001102424.1 | NP_001095894.1 | 6 - 7 | 608 | 82 | ||
NM_001102425.1 | NP_001095895.1 | 5 - 6 | 596 | 82 | ||
NM_172675.4 | NP_766263.2 | 6 - 7 | 659 | 82 | ||
GenBank mRNA | AK029571.1 | - | 7 - 8 | 649 | 82 | |
AK078110.1 | - | 5 - 6 | 659 | 82 | ||
BC094436.1 | - | 6 - 7 | 600 | 82 | ||
BC095989.1 | - | 6 - 7 | 639 | 82 | ||
BC138544.1 | - | 6 - 7 | 556 | 8861564 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |