Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007496.2 | NP_031522.2 | 6 - 7 | 4427 | 76 | |
GenBank mRNA | D26046.1 | - | 6 - 7 | 4424 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |