Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205330.1 | NP_001192259.1 | 3 - 4 | 480 | 80 | |
NM_001205332.1 | NP_001192261.1 | 3 - 4 | 480 | 80 | ||
NM_008633.4 | NP_032659.2 | 3 - 4 | 480 | 80 | ||
GenBank mRNA | BC020298.1 | - | 3 - 4 | 448 | 80 | |
BC044654.1 | - | 3 - 4 | 390 | 5400526 | 80 | |
BC050893.1 | - | 3 - 4 | 355 | 5706356 | 80 | |
BC055332.1 | - | 3 - 4 | 361 | 6407218 | 80 | |
BC055364.1 | - | 2 - 3 | 320 | 30057231 | 80 | |
M72414.1 | - | 3 - 4 | 784 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |