Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007223.3 | NP_001007224.1 | 1 - 2 | 172 | 73 | |
GenBank mRNA | BC147139.1 | - | 1 - 2 | 172 | 9056169 | 73 |
BC147140.1 | - | 1 - 2 | 172 | 9056166 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |