Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
589007 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
774635 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
262675 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
276428 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
145847 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
AK009587.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
AK150216.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
AK165627.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
BC011282.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
BC071264.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
351003 |
NM_027985.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473584 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861457 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
567080 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316912 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111884 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
61026 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123053 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P112919 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757469 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757484 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757475 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757482 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757483 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21757481 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04745591 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04745630 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112919 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123053 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04745608 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
BC116727.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK028158.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK075874.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
AK160516.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455572 |
BC116681.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137708 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112919 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123053 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01495922 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01495925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757563 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757543 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757559 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21757418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01860184 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757507 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757516 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757605 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757463 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01905138 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757485 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757500 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01306865 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757484 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757542 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757472 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757482 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757466 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01429625 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757465 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01361066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757454 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01256960 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757453 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01361060 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757496 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01157528 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757464 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02131496 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757520 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01361058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757519 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01692959 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757494 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01487633 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757604 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01306863 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757538 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757503 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01692967 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757514 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757467 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757540 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01487635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01495926 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757492 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757505 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01905129 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757606 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757489 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21757600 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01364468 |
- |
- |