Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133847.3 | NP_598608.2 | 12 - 13 | 1505 | 64 | |
GenBank mRNA | AB093220.2 | - | 11 - 12 | 1249 | 64 | |
AK035622.1 | - | 12 - 13 | 1467 | 64 | ||
AK048158.1 | - | 12 - 13 | 1263 | 64 | ||
AK083541.1 | - | 12 - 13 | 1505 | 64 | ||
AK087048.1 | - | 12 - 13 | 1258 | 64 | ||
AK088012.1 | - | 12 - 13 | 1261 | 64 | ||
AK162435.1 | - | 12 - 13 | 1266 | 64 | ||
AK171487.1 | - | 12 - 13 | 1263 | 64 | ||
AK210804.1 | - | 3 - 4 | 228 | 64 | ||
BC110309.1 | - | 12 - 13 | 1261 | 5324218 | 64 | |
BC116320.2 | - | 12 - 13 | 1457 | 40096002 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |